MilliporeSigma
  • MicroRNA-210 regulates the metabolic and inflammatory status of primary human astrocytes.
MicroRNA-210 regulates the metabolic and inflammatory status of primary human astrocytes.
Journal of neuroinflammation (2022-01-08)
Nicholas W Kieran, Rahul Suresh, Marie-France Dorion, Adam MacDonald, Manon Blain, Dingke Wen, Shih-Chieh Fuh, Fari Ryan, Roberto J Diaz, Jo Anne Stratton, Samuel K Ludwin, Joshua A Sonnen, Jack Antel, Luke M Healy
ABSTRACT
Astrocytes are the most numerous glial cell type with important roles in maintaining homeostasis and responding to diseases in the brain. Astrocyte function is subject to modulation by microRNAs (miRs), which are short nucleotide strands that regulate protein expression in a post-transcriptional manner. Understanding the miR expression profile of astrocytes in disease settings provides insight into the cellular stresses present in the microenvironment and may uncover pathways of therapeutic interest. Laser-capture microdissection was used to isolate human astrocytes surrounding stroke lesions and those from neurological control tissue. Astrocytic miR expression profiles were examined using quantitative reverse transcription polymerase chain reaction (RT-qPCR). Primary human fetal astrocytes were cultured under in vitro stress conditions and transfection of a miR mimic was used to better understand how altered levels of miR-210 affect astrocyte function. The astrocytic response to stress was studied using qPCR, enzyme-linked immunosorbent assays (ELISAs), measurement of released lactate, and Seahorse. Here, we measured miR expression levels in astrocytes around human ischemic stroke lesions and observed differential expression of miR-210 in chronic stroke astrocytes compared to astrocytes from neurological control tissue. We also identified increased expression of miR-210 in mouse white matter tissue around middle cerebral artery occlusion (MCAO) brain lesions. We aimed to understand the role of miR-210 in primary human fetal astrocytes by developing an in vitro assay of hypoxic, metabolic, and inflammatory stresses. A combination of hypoxic and inflammatory stresses was observed to upregulate miR-210 expression. Transfection with miR-210-mimic (210M) increased glycolysis, enhanced lactate export, and promoted an anti-inflammatory transcriptional and translational signature in astrocytes. Additionally, 210M transfection resulted in decreased expression of complement 3 (C3) and semaphorin 5b (Sema5b). We conclude that miR-210 expression in human astrocytes is modulated in response to ischemic stroke disease and under in vitro stress conditions, supporting a role for miR-210 in the astrocytic response to disease conditions. Further, the anti-inflammatory and pro-glycolytic impact of miR-210 on astrocytes makes it a potential candidate for further research as a neuroprotective agent.
Sigma-Aldrich
N1 Medium Supplement (100×), solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks