MilliporeSigma
  • Ultrasound-assisted surfactant-enhanced emulsification microextraction for the determination of carbamates in wines by ultra-high performance liquid chromatography-tandem mass spectrometry.
Ultrasound-assisted surfactant-enhanced emulsification microextraction for the determination of carbamates in wines by ultra-high performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2013-10-08)
David Moreno-González, José F Huertas-Pérez, Ana M García-Campaña, Juan M Bosque-Sendra, Laura Gámiz-Gracia
ABSTRACT
A new sensitive multiresidue method based on ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) has been developed for the detection, confirmation and quantification of twenty five carbamates in wine samples. The separation was achieved in 5.5 min, using a Zorbax Eclipse plus RRHD C18 column (50 mm×2.1 mm, 1.8 μm), with a mobile phase of water and methanol, both of them with 0.01% formic acid. The analytes were detected in positive mode with multiple reaction monitoring mode. Ultrasound-assisted surfactant-enhanced emulsification microextraction (UASEME), using a low-density extraction solvent has been optimized for the satisfactory extraction of carbamates and clean-up of extracts. The matrix effect was studied, showing that the proposed procedure provides very clean extracts. Under optimum conditions, recoveries for fortified wine samples ranged from 74 to 102%, with relative standard deviations lower than 6%. Limits of quantification ranged from 0.15 to 0.92 μgl(-1), showing the high sensitivity of this fast and simple method and its compliance with current requirements.
Product Description
Supelco
Ethiofencarb-sulfone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.