MilliporeSigma
  • Inhibition of aminopeptidases by amastatin and bestatin derivatives. Effect of inhibitor structure on slow-binding processes.
Inhibition of aminopeptidases by amastatin and bestatin derivatives. Effect of inhibitor structure on slow-binding processes.
Journal of medicinal chemistry (1984-04-01)
D H Rich, B J Moon, S Harbeson
ABSTRACT
Amastatin [(2S,3R)-3-amino-2-hydroxy-5-methylhexanoyl-L-valyl-L-valyl-L- aspartic acid] and bestatin [(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl-L-leucine] are slow-binding, competitive inhibitors of aminopeptidase M (AP-M) with net inhibition constants (Ki) of 1.9 X 10(-8) and 4.1 X 10(-6) M, respectively. The effect of inhibitor structure on net Ki and on slow-binding inhibition was evaluated for analogues of both inhibitors on AP-M and leucine aminopeptidase (LAP). The (2S)-hydroxyl group contributes to the stabilization of a collision complex [EI], which is formed rapidly. In contrast, increasing the peptide chain length of the inhibitor produces more potent inhibitors as a consequence of a slower binding process. A statine analogue of amastatin [(3S,4S)-Sta-Val-Val-Asp] stimulated rather than inhibited LAP. AP-M binds tri- and tetrapeptide inhibitors more strongly than dipeptide inhibitors, whereas LAP binds dipeptide inhibitors more strongly. The difference in binding can be used to distinguish cytosolic from membrane-bound aminopeptidases.