MilliporeSigma
  • Comparative glycomic profiling of isotopically permethylated N-glycans by liquid chromatography/electrospray ionization mass spectrometry.
Comparative glycomic profiling of isotopically permethylated N-glycans by liquid chromatography/electrospray ionization mass spectrometry.
Rapid communications in mass spectrometry : RCM (2013-03-16)
Yunli Hu, Janie L Desantos-Garcia, Yehia Mechref
ABSTRACT
Mass spectrometry based comparative glycomics is essential for disease biomarker discovery. However, developing a reliable quantification method is still a challenging task. We here report an isotopic labeling strategy employing stable isotopic iodomethane for comparative glycomic profiling by liquid chromatography/electrospray ionization mass spectrometry (LC/ESI-MS). N-Glycans released from model glycoproteins and blood serum samples were permethylated with iodomethane ('light') and iodomethane-d1 or -d3 ('heavy') reagents. Permethylated samples were then mixed at equal volumes prior to LC/ESI-MS analysis. Peak intensity ratios of N-glycans isotopically permethylated (Heavy/Light, H/L) were almost equal to the theoretical values. Observed differences were mainly related to the purity of 'heavy' iodomethane reagents (iodomethane-d1 or -d3). The data suggested the efficacy of this strategy to simultaneously quantify N-glycans derived from biological samples representing different cohorts. Accordingly, this strategy is effective in comparing multiple samples in a single LC/ESI-MS analysis. The potential of this strategy for defining glycomic differences in blood serum samples representing different esophageal diseases was explored. LC/ESI-MS comparative glycomic profiling of isotopically permethylated N-glycans derived from biological samples and glycoproteins reliably defined glycan changes associated with biological conditions or glycoproteins expression. As a biological application, this strategy permitted the reliable quantification of glycomic changes associated with different esophageal diseases, including high grade dysplasia, Barrett's disease, and esophageal adenocarcinoma.
Sigma-Aldrich
Iodomethane-d3, ≥99.5 atom % D, ≥99% (CP), contains copper as stabilizer