MilliporeSigma
  • Evaluation of glycolytic rates in human hematopoietic stem/progenitor cells after target gene depletion.
Evaluation of glycolytic rates in human hematopoietic stem/progenitor cells after target gene depletion.
STAR protocols (2021-06-26)
Ying Qing, Lei Gao, Li Han, Rui Su, Jianjun Chen
ABSTRACT
This protocol describes how to evaluate cellular metabolic state, including the glycolytic activity, in live CD34+ hematopoietic stem/progenitor cells (HSPCs) upon depletion of a specific target gene. We detail the procedure of enriching mononuclear cells from human umbilical cord blood samples, isolation of CD34+ HSPCs using positive immunomagnetic separation, and the analysis of glycolytic activity in lentivirally transduced CD34+ HSPCs using Seahorse Assay. This protocol can be applied to evaluate potential aerobic glycolysis gene targets for cancer therapy. For complete details on the use and execution of this protocol, please refer to Qing et al. (2021).
Sigma-Aldrich
Bovine Serum Albumin solution, 22% in saline, contains azide, aseptically filled
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture