MilliporeSigma
Role of FAP48 in HIV-associated lipodystrophy.
Journal of cellular biochemistry (2012-06-09)
Vincenzo Esposito, Lucrezia Manente, Angela Lucariello, Angelica Perna, Rosaria Viglietti, Miriam Gargiulo, Roberto Parrella, Giovanni Parrella, Alfonso Baldi, Antonio De Luca, Antonio Chirianni
ABSTRACT
The highly active antiretroviral therapy (HAART) can cause a metabolic syndrome consisting of lipodystropy/lipoatrophy, dyslipidemia, and type 2 diabetes mellitus with an increased cardiovascular risk. The pathogenetic bases of HAART-associated lipodystrophy are poorly known. A genetic screen was used to evaluate proteins that are modulated in HIV-1-infected patients with or without lipodystrophy syndrome, that are routinely treated with HAART regimens. The most significant modulation was represented by FAP48 expression. Stable over-expression of FAP48 was able to alter, in vitro, adipogenesis, acting both on calcineurin and glucocorticoid pathways. Finally, we demonstrated that FAP48 over-expression was able to influence the capacity of some HIV drugs, Saquinavir and Efavirenz, but not Stavudine, Amprenavir, and Indinavir to inhibit adipocyte formation. In conclusion, this molecule could be a potential target for novel therapeutic approaches to the HAART related lipodystrophy in HIV patients.
Sigma-Aldrich
FAP Active human, recombinant, expressed in baculovirus, ≥50% (SDS-PAGE)
Sigma-Aldrich
Saquinavir mesylate, ≥98% (HPLC), powder
Sigma-Aldrich
Amprenavir, ≥98% (HPLC)