MilliporeSigma
  • Thiazide and other Cl-benzenesulfonamide-bearing clinical drug affinities for human carbonic anhydrases.
Thiazide and other Cl-benzenesulfonamide-bearing clinical drug affinities for human carbonic anhydrases.
PloS one (2021-06-25)
Lina Baranauskiene, Lina Škiudaitė, Vilma Michailovienė, Vytautas Petrauskas, Daumantas Matulis
ABSTRACT
Twelve carbonic anhydrase (CA) isoforms catalyze carbon dioxide hydration to bicarbonate and acid protons and are responsible for many biological functions in human body. Despite their vital functions, they are also responsible for, or implicated in, numerous ailments and diseases such as glaucoma, high altitude sickness, and cancer. Because CA isoforms are highly homologous, clinical drugs designed to inhibit enzymatic activity of a particular isoform, can also bind to others with similar affinity causing toxic side effects. In this study, the affinities of twelve CA isoforms have been determined for nineteen clinically used drugs used to treat hypertension related diseases, i.e. thiazides, indapamide, and metolazone. Their affinities were determined using a fluorescent thermal shift assay. Stopped flow assay and isothermal titration calorimetry were also employed on a subset of compounds and proteins to confirm inhibition of CA enzymatic activity and verify the quantitative agreement between different assays. The findings of this study showed that pharmaceuticals could bind to human CA isoforms with variable affinities and inhibit their catalytic activity, even though the drug was intended to interact with a different (non-CA) protein target. Relatively minor structural changes of the compounds may cause significant changes in affinity and selectivity for a particular CA isoform.
Product Description
Altizide, European Pharmacopoeia (EP) Reference Standard