MilliporeSigma
  • Colistin Treatment Affects Lipid Composition of Acinetobacter baumannii.
Colistin Treatment Affects Lipid Composition of Acinetobacter baumannii.
Antibiotics (Basel, Switzerland) (2021-06-03)
Ye Tao, Sébastien Acket, Emma Beaumont, Henri Galez, Luminita Duma, Yannick Rossez
ABSTRACT
Multidrug-resistant Acinetobacter baumannii (A. baumannii) causes severe and often fatal healthcare-associated infections due partly to antibiotic resistance. There are no studies on A. baumannii lipidomics of susceptible and resistant strains grown at lethal and sublethal concentrations. Therefore, we analyzed the impact of colistin resistance on glycerolipids' content by using untargeted lipidomics on clinical isolate. Nine lipid sub-classes were annotated, including phosphatidylcholine, rarely detected in the bacterial membrane among 130 different lipid species. The other lipid sub-classes detected are phosphatidylethanolamine (PE), phosphatidylglycerol (PG), lysophosphatidylethanolamine, hemibismonoacylglycerophosphate, cardiolipin, monolysocardiolipin, diacylglycerol, and triacylglycerol. Under lethal and sublethal concentrations of colistin, significant reduction of PE was observed on the resistant and susceptible strain, respectively. Palmitic acid percentage was higher at colistin at low concentration but only for the susceptible strain. When looking at individual lipid species, the most abundant PE and PG species (PE 34:1 and PG 34:1) are significantly upregulated when the susceptible and the resistant strains are cultivated with colistin. This is, to date, the most exhaustive lipidomics data compilation of A. baumannii cultivated in the presence of colistin. This work is highlighting the plasma membrane plasticity used by this gram-negative bacterium to survive colistin treatment.
Product Description
Supelco
Colistin Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material