MilliporeSigma
  • Delta opioid receptors expressed by stably transfected jurkat cells signal through the map kinase pathway in a ras-independent manner.
Delta opioid receptors expressed by stably transfected jurkat cells signal through the map kinase pathway in a ras-independent manner.
Journal of neuroimmunology (1999-06-22)
N A Shahabi, Y Daaka, K McAllen, B M Sharp
ABSTRACT
Delta opioid receptors (DOR) are G-protein coupled 7-transmembrane receptors (GPCR), expressed by thymic and splenic T cells, that modulate interleukin (IL)-2 production and proliferation in response to concanavalin A or crosslinking the TCR. Mitogen-activated protein kinases (MAPKs) are involved in mediating intracellular responses to TCR crosslinking. In addition, MAPKs can be activated by signaling cascades that are initiated by the release of G-proteins from GPCRs. To determine whether DORs expressed by T cells signal through the MAPKs, extracellular-regulated kinases (ERKs) 1 and 2, two delta opioid peptides, deltorphin and [D-Ala2,D-Leu5]-enkephalin (DADLE), were studied in Jurkat cells that had been stably transfected with DOR (DOR-Ju.1). These peptides rapidly and dose-dependently induced ERK phosphorylation; pretreatment with naltrindole (NTI), a selective DOR antagonist, abolished this. Pertussis toxin (PTX) also inhibited phosphorylation, indicating the involvement of the Gi/o proteins. Herbimycin A, a protein tyrosine kinase (PTK) inhibitor, reduced the DADLE-induced ERK phosphorylation by 68%. ERK phosphorylation was inhibited by Bisindolylmaleimide 1 (GF109203X), an inhibitor of PKC, and by pretreatment with PMA prior to DADLE. A GTP/GDP exchange assay was used to assess the potential role of Ras in the pathway leading to ERK phosphorylation; DADLE failed to stimulate GTP/GDP exchange in comparison to PMA. Additional studies showed that DADLE stimulated an increase in cfos mRNA; this was reduced by the inhibitor of MAPK/ERK kinase (MEK), PD98059. Therefore, in DOR-Ju.1 cells, DOR agonists stimulate ERK phosphorylation in a Ras independent and PKC-dependent manner; PTKs appear to be involved. MAPKs mediate the increase in cfos mRNA induced by DOR agonists.
Sigma-Aldrich
[D-Ala2, D-Leu5]-Enkephalin acetate salt, ≥95% (HPLC)