MilliporeSigma
Apolipoprotein E derived from CD11c+ cells ameliorates atherosclerosis.
iScience (2022-01-18)
Manuela Sauter, Reinhard J Sauter, Henry Nording, Chaolan Lin, Marcus Olbrich, Stella Autenrieth, Christian Gleissner, Martin Thunemann, Nadia Otero, Esther Lutgens, Zouhair Aherrahrou, Dennis Wolf, Lars Zender, Sven Meuth, Robert Feil, Harald F Langer
ABSTRACT
Atherosclerosis is studied in models with dysfunctional lipid homeostasis-predominantly the ApoE-/- mouse. The role of antigen-presenting cells (APCs) for lipid homeostasis is not clear. Using a LacZ reporter mouse, we showed that CD11c+ cells were enriched in aortae of ApoE-/- mice. Systemic long-term depletion of CD11c+ cells in ApoE-/- mice resulted in significantly increased plaque formation associated with reduced serum ApoE levels. In CD11ccre+ApoEfl/fl and Albumincre+ApoEfl/fl mice, we could show that ≈70% of ApoE is liver-derived and ≈25% originates from CD11c+ cells associated with significantly increased atherosclerotic plaque burden in both strains. Exposure to acLDL promoted cholesterol efflux from CD11c+ cells and cell-specific deletion of ApoE resulted in increased inflammation reflected by increased IL-1β serum levels. Our results determined for the first time the level of ApoE originating from CD11c+ cells and demonstrated that CD11c+ cells ameliorate atherosclerosis by the secretion of ApoE.
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Roche
Streptavidin-AP conjugate, solution, pkg of 1 mL (1,000U)