MilliporeSigma
  • ALKBH5-mediated m6A modification of lncRNA KCNQ1OT1 triggers the development of LSCC via upregulation of HOXA9.
ALKBH5-mediated m6A modification of lncRNA KCNQ1OT1 triggers the development of LSCC via upregulation of HOXA9.
Journal of cellular and molecular medicine (2021-12-02)
Yushan Li, Bingrui Yan, Xin Wang, Qiuying Li, Xuan Kan, Jingting Wang, Yanan Sun, Peng Wang, Linli Tian, Ming Liu
ABSTRACT
It has been shown that N6-methyladenosine (m6A) modification is involved in the development of complex human diseases, especially in the development of cancer. Our research investigated the role and mechanism of the m6A modification of lncRNA KCNQ1 overlapping transcript 1 (KCNQ1OT1) in Laryngeal squamous cell carcinoma (LSCC) progression. Microarray analysis was used to quantitatively detect the m6A apparent transcriptional modification level of lncRNA in LSCC tissue. Methylated RNA immunoprecipitation-qPCR (MeRIP-qPCR), in situ hybridization (ISH) and quantitative real-time PCR (qRT-PCR) were used to examine the m6A modification and expression of KCNQ1OT1. In addition, in vivo and in vitro experiments have tested the effects of KCNQ1OT1 knockdown on the proliferation, invasion and metastasis of LSCC. Mechanically, we found the N6-methyladenosine (m6A) demethylase ALKBH5 mediates KCNQ1OT1 expression via an m6A-YTHDF2-dependent manner and KCNQ1OT1 could directly bind to HOXA9 to further regulate the proliferation, invasion and metastasis of LSCC cells. In general, our research indicates that ALKBH5-mediated m6A modification of KCNQ1OT1 triggers the development of LSCC via upregulation of HOXA9.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.