MilliporeSigma
Prevention of oxidative DNA degradation by copper-binding peptides.
Bioscience, biotechnology, and biochemistry (2011-07-09)
Ken Yokawa, Tomoko Kagenishi, Tomonori Kawano
ABSTRACT
Free divalent ions of copper (Cu) are capable of generating radical species such as hydroxyl radicals in the presence of hydrogen peroxide or ascorbic acid through Harbor-Weiss-like reactions under physiological conditions. It has been reported that radical-mediated damage to DNA molecules in animal cells leads to programmed cell death. Hence it is important to seek for methods to prevent Cu-mediated DNA damage. In this study we identified on effect of Cu binding of short peptides (chiefly Gly-Gly-His tripeptide) in the prevention of DNA degradation caused by Cu-mediated reactions in the presence of hydrogen peroxide and of ascorbic acid.
Sigma-Aldrich
Gly-Gly-His