MilliporeSigma
In vitro detoxification of cyclosarin (GF) by modified cyclodextrins.
Toxicology letters (2010-11-03)
Susanne Müller, Marianne Koller, Romain Le Provost, Olivier Lafont, François Estour, Timo Wille, Horst Thiermann, Franz Worek, Georg Reiter
ABSTRACT
Developing potent detoxification strategies for prophylaxis and therapy against organophosphate (OP) intoxication still represents a challenging task. Clinical application of numerous investigated substances including enzymes and low molecular scavengers like metal ions or nucleophiles could not yet be realised due to profound disadvantages. Presenting a promising attempt, cyclodextrins (CDs) efficiently enhance the degradation of some organophosphorus compounds. The present study examined the in vitro GF degradation mediated by three CDs and a nucleophilic precursor performed by mass spectrometric detection with ammonia chemical ionisation. All four compounds caused a notable enhancement of GF detoxification that was synergistically accelerated in the case of 2-O-(3-carboxy-4-iodosobenzyl)-β-cyclodextrin (IBA-β-CD) with the alpha-nucleophile 2-iodosobenzoic acid (IBA) grafted on the secondary face of β-cyclodextrin (β-CD). In vitro toxicokinetic investigations of CD derivatives are needed to evaluate the effect of slow terminal elimination phase of the more toxic (-)-GF shown for two CD-derivatives underlining the necessity of detecting the complete kinetic course of inactivation. The observed effect of fast high affinity binding (20-30%) represents an additional therapeutic option of an extremely rapid reduction of GF concentration in vivo. Distinctive differences in the course of reaction are detected depending on β-CD-derivatives, allowing a first inference of possible mechanisms and relevance of attached substituents. However, further profound investigation needs to be done to evaluate the basis of a clinical application of substituted CDs as potential detoxification agents.
Sigma-Aldrich
2-Iodosobenzoic acid