MilliporeSigma
  • Interleukin-10 and immunity against prokaryotic and eukaryotic intracellular pathogens.
Interleukin-10 and immunity against prokaryotic and eukaryotic intracellular pathogens.
Infection and immunity (2011-05-18)
Joshua C Cyktor, Joanne Turner
ABSTRACT
The generation of an effective immune response against an infection while also limiting tissue damage requires a delicate balance between pro- and anti-inflammatory responses. Interleukin-10 (IL-10) has potent immunosuppressive effects and is essential for regulation of immune responses. However, the immunosuppressive properties of IL-10 can also be exploited by pathogens to facilitate their own survival. In this minireview, we discuss the role of IL-10 in modulating intracellular bacterial, fungal, and parasitic infections. Using information from several different infection models, we bring together and highlight some common pathways for IL-10 regulation and function that cannot be fully appreciated by studies of a single pathogen.