MilliporeSigma
  • Characterization of an APP/tau rat model of Alzheimer's disease by positron emission tomography and immunofluorescent labeling.
Characterization of an APP/tau rat model of Alzheimer's disease by positron emission tomography and immunofluorescent labeling.
Alzheimer's research & therapy (2021-10-18)
Thomas Filip, Severin Mairinger, Joerg Neddens, Michael Sauberer, Stefanie Flunkert, Johann Stanek, Thomas Wanek, Nobuyuki Okamura, Oliver Langer, Birgit Hutter-Paier, Claudia Kuntner
ABSTRACT
To better understand the etiology and pathomechanisms of Alzheimer's disease, several transgenic animal models that overexpress human tau or human amyloid-beta (Aβ) have been developed. In the present study, we generated a novel transgenic rat model by cross-breeding amyloid precursor protein (APP) rats with tau rats. We characterized this model by performing positron emission tomography scans combined with immunofluorescent labeling and cerebrospinal fluid analyses. APP/Tau rats were generated by cross-breeding male McGill-R-Thy1-APP transgenic rats with female hTau-40/P301L transgenic rats. APP/Tau double transgenic rats and non-transgenic (ntg) littermates aged 7, 13, and 21 months were subjected to dynamic [11C] PiB scan and dynamic [18F]THK-5317 scans. For regional brain analysis, a template was generated from anatomical MR images of selected animals, which was co-registered with the PET images. Regional analysis was performed by application of the simplified reference tissue model ([11C]PiB data), whereas [18F]THK-5317 data were analyzed using a 2-tissue compartment model and Logan graphical analysis. In addition, immunofluorescent labeling (tau, amyloid) and cerebrospinal fluid analyses were performed. [11C]PiB binding potential (BPND) and [18F]THK-5317 volume of distribution (VT) showed an increase with age in several brain regions in the APP/Tau group but not in the ntg control group. Immunohistochemical analysis of brain slices of PET-scanned animals revealed a positive correlation between Aβ labeling and [11C]PiB regional BPND. Tau staining yielded a trend towards higher levels in the cortex and hippocampus of APP/Tau rats compared with ntg littermates, but without reaching statistical significance. No correlation was found between tau immunofluorescence labeling results and the respective [18F]THK-5317 VT values. We thoroughly characterized a novel APP/Tau rat model using combined PET imaging and immunofluorescence analysis. We observed an age-related increase in [11C]PiB and [18F]THK-5317 binding in several brain regions in the APP/Tau group but not in the ntg group. Although we were able to reveal a positive correlation between amyloid labeling and [11C]PiB regional brain uptake, we observed relatively low human tau and amyloid fibril expression levels and a somewhat unstable brain pathology which questions the utility of this animal model for further studies.
Sigma-Aldrich
Anti-Amyloid Fibrils LOC Antibody, serum, Chemicon®