MilliporeSigma
  • Quantification of plasma lactate concentrations using capillary electrophoresis with contactless conductivity detection.
Quantification of plasma lactate concentrations using capillary electrophoresis with contactless conductivity detection.
Electrophoresis (2011-03-12)
Worapan Pormsila, Réjane Morand, Stephan Krähenbühl, Peter C Hauser
ABSTRACT
The quantification of plasma lactate and evaluation of the lactate threshold by CE with capacitively coupled contactless conductivity is demonstrated. The only sample preparation needed was deproteinization with a ACN/methanol mixture. A solution of 10 mmol/L 2-morpholinoethanesulfonic acid monohydrate, 10 mmol/L DL-histidine, 70 μmol/L hexadecyltrimethylammonium bromide, pH 6.0 was found suitable as running buffer. Linearity was achieved for the concentration range of 10-1000 μmol/L with a correlation coefficient of 0.9994. The limit of detection (3 S/N) was determined as 3.2 μmol/L. Intra- and inter-day variabilities were less than 7% RSD. The suitability of the method could be demonstrated by analyzing various clinical samples, where the results correlated satisfactorily with those of an established enzymatic method.
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)