MilliporeSigma
  • Data-independent liquid chromatography/mass spectrometry (LC/MS(E)) detection and quantification of the secreted Apium graveolens pathogen defense protein mannitol dehydrogenase.
Data-independent liquid chromatography/mass spectrometry (LC/MS(E)) detection and quantification of the secreted Apium graveolens pathogen defense protein mannitol dehydrogenase.
Rapid communications in mass spectrometry : RCM (2010-03-10)
Kevin Blackburn, Fang-Yi Cheng, John D Williamson, Michael B Goshe
ABSTRACT
Plant cells secrete a wide variety of defense-related proteins into the extracellular space or apoplast in response to pathogen attack. One of these, mannitol dehydrogenase (MTD), is normally a cytoplasmic enzyme whose primary role is the regulation of intracellular levels of the sugar alcohol mannitol in plants. Recent immunological and biochemical evidence, however, suggests that MTD is also secreted into the apoplast in response to pathogen attack, despite lacking a known peptide signal sequence for Golgi-mediated secretion. Because many plant pathogenic fungi secrete mannitol to overcome pathogen-induced generation of reactive oxygen species (ROS) by the plant, extracellular localization of MTD is hypothesized to have a defensive role of catabolizing pathogen-secreted mannitol. In the current study, LC/MS(E) was used to analyze proteins in the secretome of Apium graveolens (celery) following treatment with salicylic acid (SA), an endogenous elicitor of defense responses in plants. Levels of MTD in the secretome of SA-treated celery cell cultures were found to be induced at least 18-fold over secretome samples from cell cultures not exposed to SA. This value is in close agreement with published immunological and biochemical observations. Overall, this report provides the first mass spectrometry identification and quantification measurements supporting the hypothesis that MTD is secreted in response to simulated pathogen attack via a non-classical secretion mechanism. As demonstrated with MTD secretion, LC/MS(E) can be implemented as a discovery-driven MRM-based quantitative approach which can be used to reveal potential post-translational modifications, thus providing a new method in the area of gel-free and label-free proteomic analysis.
Sigma-Aldrich
Mannitol Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, ≥60 units/mg protein