MilliporeSigma
  • Cargo-specific recruitment in clathrin- and dynamin-independent endocytosis.
Cargo-specific recruitment in clathrin- and dynamin-independent endocytosis.
Nature cell biology (2021-10-08)
Paulina Moreno-Layseca, Niklas Z Jäntti, Rashmi Godbole, Christian Sommer, Guillaume Jacquemet, Hussein Al-Akhrass, James R W Conway, Pauliina Kronqvist, Roosa E Kallionpää, Leticia Oliveira-Ferrer, Pasquale Cervero, Stefan Linder, Martin Aepfelbacher, Henrik Zauber, James Rae, Robert G Parton, Andrea Disanza, Giorgio Scita, Satyajit Mayor, Matthias Selbach, Stefan Veltel, Johanna Ivaska
ABSTRACT
Spatially controlled, cargo-specific endocytosis is essential for development, tissue homeostasis and cancer invasion. Unlike cargo-specific clathrin-mediated endocytosis, the clathrin- and dynamin-independent endocytic pathway (CLIC-GEEC, CG pathway) is considered a bulk internalization route for the fluid phase, glycosylated membrane proteins and lipids. While the core molecular players of CG-endocytosis have been recently defined, evidence of cargo-specific adaptors or selective uptake of proteins for the pathway are lacking. Here we identify the actin-binding protein Swiprosin-1 (Swip1, EFHD2) as a cargo-specific adaptor for CG-endocytosis. Swip1 couples active Rab21-associated integrins with key components of the CG-endocytic machinery-Arf1, IRSp53 and actin-and is critical for integrin endocytosis. Through this function, Swip1 supports integrin-dependent cancer-cell migration and invasion, and is a negative prognostic marker in breast cancer. Our results demonstrate a previously unknown cargo selectivity for the CG pathway and a role for specific adaptors in recruitment into this endocytic route.
Sigma-Aldrich
Laminin from human fibroblasts, cell culture derived, liquid, sterile-filtered
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture
Sigma-Aldrich
Collagen Type I, rat tail
Sigma-Aldrich
Monoclonal Anti-HLA Class I, low endotoxin antibody produced in mouse, clone W6/32, purified immunoglobulin, buffered aqueous solution