MilliporeSigma
  • FSC231 alleviates paclitaxel-induced neuralgia by inhibiting the interactions between PICK1 and GluA2 and activates GSK-3β and ERK1/2.
FSC231 alleviates paclitaxel-induced neuralgia by inhibiting the interactions between PICK1 and GluA2 and activates GSK-3β and ERK1/2.
Brain and behavior (2021-09-29)
Xi Zhang, Jiagao Wang, Ran Ran, Yuchuan Peng, Yun Xiao
ABSTRACT
FSC231, a PSD-95/DLG/ZO-1 (PDZ) domain inhibitor of protein kinase Cα interacting protein 1 (PICK1), has analgesic effects, but the mechanism remains unclear. The expression level of PICK1 in dorsal root ganglion (DRG) of rats was changed by vector plasmid, and the effect of PICK1 on paclitaxel (PTL)-induced neuralgia of rats was observed in collaboration with FSC231 treatment. The possible molecular mechanisms were explored by quantitative real-time polymerase chain reaction (qRT-PCR), Western Blot and co-immunoprecipitation (Co-IP) techniques. PTL treatment can significantly reduce mechanical withdrawal threshold (MWT), shorten thermal withdrawal latency (TWL), promote DRG inflammation and release of substance P (SP), stimulate PICK1 expression, decrease α-amino-3-hydroxy-5-methyl-4-isoxazole-propionic acid receptor 2 (AMPAR, GluA2) level and increase glycogen synthase kinase-3β (GSK-3β) and extracellular regulated protein kinases1/2 (ERK1/2) phosphorylation in rats, while FSC231 treatment can alleviate the above effects induced by PTL. Overexpression of PICK1 can counteract reduced PICK1 level, increased GluA2 level and decreased GSK-3β and ERK1/2 phosphorylation levels caused by FSC231 treatment. The results of Co-IP confirmed the interactions between PICK1 and GluA2. Both FSC231 treatment and silent PICK1 improved PTL-induced MWT reduction, TWL shortening, inflammation, SP release and related gene expression changes, with cumulative effect. FSC231 activates GSK-3β/ERK1/2 by inhibiting the interaction between PICK1 and GluA2 and alleviates PTL-induced DRG neuralgia in rats.
Sigma-Aldrich
Anti-Substance P antibody produced in rabbit, whole antiserum