MilliporeSigma
  • LRIK interacts with the Ku70-Ku80 heterodimer enhancing the efficiency of NHEJ repair.
LRIK interacts with the Ku70-Ku80 heterodimer enhancing the efficiency of NHEJ repair.
Cell death and differentiation (2020-06-27)
Dan Wang, Zheng Zhou, Erzhong Wu, Can Ouyang, Guifeng Wei, Yunfei Wang, Dandan He, Ya Cui, Dongdong Zhang, Xiaomin Chen, Simon H Reed, Jianjun Luo, Runsheng Chen
ABSTRACT
Despite recent advances in our understanding of the function of long noncoding RNAs (lncRNAs), their roles and functions in DNA repair pathways remain poorly understood. By screening a panel of uncharacterized lncRNAs to identify those whose transcription is induced by double-strand breaks (DSBs), we identified a novel lncRNA referred to as LRIK that interacts with Ku, which enhances the ability of the Ku heterodimer to detect the presence of DSBs. Here, we show that depletion of LRIK generates significantly enhanced sensitivity to DSB-inducing agents and reduced DSB repair efficiency. In response to DSBs, LRIK enhances the recruitment of repair factors at DSB sites and facilitates γH2AX signaling. Our results demonstrate that LRIK is necessary for efficient repairing DSBs via nonhomologous end-joining pathway.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Rabbit anti-Ku80 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.