MilliporeSigma
  • From IL-10 to IL-12: how pathogens and their products stimulate APCs to induce T(H)1 development.
From IL-10 to IL-12: how pathogens and their products stimulate APCs to induce T(H)1 development.
Nature immunology (2009-08-21)
Anne O'Garra, Kenneth M Murphy
ABSTRACT
The authors recount their discovery of how pathogen-induced interleukin 12 production leads to T(H)1 T cell polarization. Simultaneously they discovered the suppressive cytokine interleukin 10 inhibits antigen-presenting cells, thus regulating development of T(H)1 cells.