MilliporeSigma
  • Effects of Selenium Supplement on B Lymphocyte Activity in Experimental Autoimmune Thyroiditis Rats.
Effects of Selenium Supplement on B Lymphocyte Activity in Experimental Autoimmune Thyroiditis Rats.
International journal of endocrinology (2021-08-24)
Yang Li, Xinhe Zuo, Chuan Hua, Yong Zhao, Xun Pei, Man Tian
ABSTRACT
45 healthy and adult female SD rats were randomly divided into three groups: normal control group, EAT model group, and selenium yeast supplement EAT group. The EAT model rats were induced by subcutaneous injection of porcine thyroglobulin and fed with high iodine water. The concentrations of serum thyroid-stimulating hormone (TSH), TGAb, TPOAb, and B cell activating factor (BAFF) were detected in each group by enzyme-linked immunosorbent assay (ELISA), and the expression of interleukin-10 (IL-10) in thyroid tissue was detected by immunohistochemistry. B cells and regulatory B cells (Bregs) ratios in the spleen of rats were analyzed by flow cytometry. In contrast with the EAT model group, the levels of serum TSH, TGAB, TPOAb, and BAFF were decreased, while IL-10 expression was increased in thyroid tissue, and Bregs ratio was upregulated in the spleen (all p < 0.05) in the selenium yeast supplement EAT group. Selenium yeast supplement could partially attenuate immune imbalance in EAT rats, which may be related to the mechanism of modulating B lymphocyte activity.
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder