MilliporeSigma
  • Sex differences in visceral fat lipolysis and metabolic complications of obesity.
Sex differences in visceral fat lipolysis and metabolic complications of obesity.
Arteriosclerosis, thrombosis, and vascular biology (1997-07-01)
F Lönnqvist, A Thörne, V Large, P Arner
ABSTRACT
Cardiovascular complications of obesity are more common in men than women. Sex differences in visceral fat lipolysis may be of importance in this respect, since increased release of free fatty acids (FFAs) from visceral fat to the liver by the portal venous system has been thought to cause several metabolic complications due to obesity, such as hypertension, hyperlipidemia, and glucose intolerance. The aim of this study was to investigate sex differences in clinical characteristics and visceral fat mobilization in obesity. Obese subjects (22 male and 23 female) undergoing elective surgery were matched for body mass index and age. The males had both higher waist-to-hip ratio (WHR), sagittal diameter, blood pressure, fat-cell volume, plasma insulin, glucose, and triglyceride and lower HDL cholesterol levels than the females. The rate of norepinephrine-induced FFA and glycerol release was twofold higher in men (P = .02). No significant reutilization of FFA was observed. The difference in maximum norepinephrine-induced rate of lipolysis between men and women was independent of both WHR and sagittal diameter and was an independent regressor for levels of plasma glucose and plasma HDL cholesterol. Fat-cell volume was an independent regressor for plasma triglycerides and blood pressure. No sex differences in the lipolytic sensitivity to beta 1- or beta 2-adrenoceptor-specific agonists or in the antilipolytic effect of insulin were observed. However, the lipolytic beta 3-adrenoceptor sensitivity was 12 times higher (P = .004) and the antilipolytic alpha 2-adrenoceptor sensitivity 17 times lower (P = .003) in men. Furthermore, lipolysis induced by agents acting at the adenylate cyclase and protein kinase A levels were almost twofold enhanced in men. However, no sex difference in maximum hormone-sensitive lipase activity was observed. In conclusion, in obesity, catecholamine-induced rate of FFA mobilization from visceral fat to the portal venous system is higher in men than women. This phenomenon is partly due to a larger fat-cell volume but also to a decrease in the function of alpha 2-adrenoceptors, an increase in the function of beta 3-adrenoceptors, and an increased ability of cyclic AMP to activate hormone-sensitive lipase. These factors may contribute to gender-specific differences in metabolic and cardiovascular disturbances accompanied by obesity.
Sigma-Aldrich
Glycerokinase from Escherichia coli, lyophilized powder, 40-100 units/mg protein