MilliporeSigma
Zero-preserving imputation of single-cell RNA-seq data.
Nature communications (2022-01-13)
George C Linderman, Jun Zhao, Manolis Roulis, Piotr Bielecki, Richard A Flavell, Boaz Nadler, Yuval Kluger
ABSTRACT
A key challenge in analyzing single cell RNA-sequencing data is the large number of false zeros, where genes actually expressed in a given cell are incorrectly measured as unexpressed. We present a method based on low-rank matrix approximation which imputes these values while preserving biologically non-expressed genes (true biological zeros) at zero expression levels. We provide theoretical justification for this denoising approach and demonstrate its advantages relative to other methods on simulated and biological datasets.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Dispase® II, protease