MilliporeSigma
  • Parkinson's Disease-Related Biomarkers That May Appear in Amphetamine Abusers.
Parkinson's Disease-Related Biomarkers That May Appear in Amphetamine Abusers.
BioMed research international (2021-10-30)
Aziza Al-Rafiah, Rania Magadmi, AbdulAziz Al-Kaabi, Nimah Alsomali
ABSTRACT
Parkinson's disease (PD) is one of the most common neurodegenerative disorders. Amphetamine addiction may cause serious of psychotic and physical damage to humans. There is some evidence that shows that amphetamine may increase the risk of PD. Thus, this study is aimed at comparing the PD serum biomarkers between amphetamine addicts and PD patients and utilizing them as diagnostic biomarkers for the early detection of PD incidence among amphetamine addicts. In the current study, nineteen amphetamine addicts, aged <40, were recruited from the Al Amal Psychiatric hospital, Jazan, Saudi Arabia. Nineteen PD patients and 19 healthy controls, who have never taken amphetamine, were also recruited. Blood samples were withdrawn from all groups. A biomarker multiplex assay from MILLIPLEX was used to assess the levels of serum amyloid-P (SAP), complement C4, C-reactive protein (CRP), and CRP/albumin ratio in serum samples (Vitros 350® slide was used to assess the albumin). All data were statistically analyzed using one-way ANOVA. The results showed that SAP and CRP levels were significantly higher in amphetamine addicts compared to healthy controls (p = 0.0001 and p = 0.0001, respectively). The results of amphetamine addicts were comparable to PD levels. However, there are no significant differences between all studied groups concerning complement C4 level. Moreover, albumin levels were significantly decreased and CRP/Albumin ratio levels were significantly increased in amphetamine addicts (p = 0.01 and p = 0.041, respectively) in contrast with controls. These findings indicate that the increased level of these inflammatory biomarkers (SAP and CRP) in the amphetamine addicts may give a potential possibility of their serum level to be used as screening markers to detect PD development in the amphetamine addict. It may be useful to evaluate the changes in easily accessible and cost-effective parameters such as the serum CRP/albumin ratio.
Product Description
Millipore
MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 2 - Neuroscience Multiplex Assay, The analytes available for this multiplex kit are: α1-Antitrypsin, Complement C4, CRP, MIP-4, PEDF, SAP.