MilliporeSigma
Inhibitory role of endophilin 3 in receptor-mediated endocytosis.
The Journal of biological chemistry (2004-04-07)
Hiroko Sugiura, Ken Iwata, Masato Matsuoka, Hiroshi Hayashi, Takako Takemiya, Shin Yasuda, Masumi Ichikawa, Takashi Yamauchi, Patrick Mehlen, Tatsuya Haga, Kanato Yamagata
ABSTRACT
Endophilin 1 (Endo1) participates in synaptic vesicle biogenesis through interactions of its Src homology 3 domain with the polyphosphoinositide phosphatase Synaptojanin and the GTPase Dynamin. Endo1 has also been reported to affect endocytosis by converting membrane curvature via its lysophosphatidic acid acyltransferase activity. Here we report that a closely related isoform of Endo1, Endo3, inhibits clathrin-mediated endocytosis. Mutational analyses showed that the variable region of Endo3 is important in regulating transferrin endocytosis. In the brain, Endo3 is co-localized with dopamine D2 receptor in olfactory nerve terminals and inhibits its clathrin-mediated endocytosis in COS-7 cells. Furthermore, overexpression of Endo3 in an olfactory epithelium-derived cell line suppressed dopamine D2 receptor-mediated endocytosis and therefore accelerated its dopamine-induced differentiation. These results indicate that Endo3 may act as a negative regulator of clathrin-mediated endocytosis in brain neurons.
Sigma-Aldrich
Monoclonal Anti-CD71-Biotin antibody produced in mouse, clone MEM-75, purified immunoglobulin, buffered aqueous solution