MilliporeSigma
  • Influence of Human Telomerase Reverse Transcriptase Mutation on the Aggressiveness and Recurrence in Meningiomas.
Influence of Human Telomerase Reverse Transcriptase Mutation on the Aggressiveness and Recurrence in Meningiomas.
Cureus (2021-07-09)
Balkan Sahin, Salim Katar, Saime A Şahin, Serdar Çevik, Sevket Evran, Oguz Baran, Canan Tanık, Hüseyin U Adılay, Adem Yılmaz
ABSTRACT
Over 200 human telomerase reverse transcriptase (hTERT) polymorphism combinations have been implicated in the development of cancer. This study aimed to evaluate hTERT mutations in meningioma tissue and its association with meningioma. A total of 90 patients who underwent surgery between 2006 and 2015 and were histopathologically diagnosed with meningioma (WHO 2016) were included. Among the 90 participants included herein, 50 (55.5%) and 40 (44.5%) were female and male, respectively, with an average age of 56.2 ± 14 years. Mean Ki-67 values were 10.56% (SD 12.41, range 0-60), while the mean follow-up duration was 39.1 months (SD 26.3). Low- and high-grade patients had a mean Ki-67 score of 4.31% (SD 3.58, range 0-16) and 19.92% (SD 14.91, range 2-60) (p = 0.0001). Our results showed a moderate positive correlation between Ki-67 score and the presence of hTERT mutation (Pearson correlation test, r = 0.5161; p = 0.0001). Patients with an hTERT mutation > 30% had significantly higher risk for reoperation than those with lower levels of mutation (p = 0.016, chi square test). None of the patients requiring reoperation had an hTERT mutation < 10%. Moreover, high-grade patients had a 7.2 times higher risk of reoperation than those with an hTERT mutation > 30%. The presence of hTERT mutation, in addition to high Ki-67, indicated a more aggressive meningioma disease course and potentially increased risk of recurrence.
Sigma-Aldrich
Anti-TERT Antibody, from rabbit, purified by affinity chromatography