MilliporeSigma
  • Time-restricted feeding prevents deleterious metabolic effects of circadian disruption through epigenetic control of β cell function.
Time-restricted feeding prevents deleterious metabolic effects of circadian disruption through epigenetic control of β cell function.
Science advances (2021-12-16)
Matthew R Brown, Satish K Sen, Amelia Mazzone, Tracy K Her, Yuning Xiong, Jeong-Heon Lee, Naureen Javeed, Christopher S Colwell, Kuntol Rakshit, Nathan K LeBrasseur, Alexandre Gaspar-Maia, Tamas Ordog, Aleksey V Matveyenko
ABSTRACT
Circadian rhythm disruption (CD) is associated with impaired glucose homeostasis and type 2 diabetes mellitus (T2DM). While the link between CD and T2DM remains unclear, there is accumulating evidence that disruption of fasting/feeding cycles mediates metabolic dysfunction. Here, we used an approach encompassing analysis of behavioral, physiological, transcriptomic, and epigenomic effects of CD and consequences of restoring fasting/feeding cycles through time-restricted feeding (tRF) in mice. Results show that CD perturbs glucose homeostasis through disruption of pancreatic β cell function and loss of circadian transcriptional and epigenetic identity. In contrast, restoration of fasting/feeding cycle prevented CD-mediated dysfunction by reestablishing circadian regulation of glucose tolerance, β cell function, transcriptional profile, and reestablishment of proline and acidic amino acid–rich basic leucine zipper (PAR bZIP) transcription factor DBP expression/activity. This study provides mechanistic insights into circadian regulation of β cell function and corresponding beneficial effects of tRF in prevention of T2DM.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Foxo1 Inhibitor, AS1842856, Foxo1 Inhibitor, AS1842856, is a cell-permeable inhibitor that blocks the transcription activity of Foxo1 (IC₅₀ = 33 nM). Directly binds to the active Foxo1, but not the Ser256-phosphorylated form.