MilliporeSigma
  • The SNAG Domain of Insm1 Regulates Pancreatic Endocrine Cell Differentiation and Represses β- to δ-Cell Transdifferentiation.
The SNAG Domain of Insm1 Regulates Pancreatic Endocrine Cell Differentiation and Represses β- to δ-Cell Transdifferentiation.
Diabetes (2021-02-07)
Xuehua Liang, Hualin Duan, Yahui Mao, Ulrich Koestner, Yiqiu Wei, Feng Deng, Jingshen Zhuang, Huimin Li, Cunchuan Wang, Luis R Hernandez-Miranda, Weihua Tao, Shiqi Jia
ABSTRACT
The allocation and specification of pancreatic endocrine lineages are tightly regulated by transcription factors. Disturbances in differentiation of these lineages contribute to the development of various metabolic diseases, including diabetes. The insulinoma-associated protein 1 (Insm1), which encodes a protein containing one SNAG domain and five zinc fingers, plays essential roles in pancreatic endocrine cell differentiation and in mature β-cell function. In the current study, we compared the differentiation of pancreatic endocrine cells between Insm1 null and Insm1 SNAG domain mutants (Insm1delSNAG) to explore the specific function of the SNAG domain of Insm1. We show that the δ-cell number is increased in Insm1delSNAG but not in Insm1 null mutants as compared with the control mice. We also show a less severe reduction of the β-cell number in Insm1delSNAG as that in Insm1 null mutants. In addition, similar deficits are observed in α-, PP, and ε-cells in Insm1delSNAG and Insm1 null mutants. We further identified that the increased δ-cell number is due to β- to δ-cell transdifferentiation. Mechanistically, the SNAG domain of Insm1 interacts with Lsd1, the demethylase of H3K4me1/2. Mutation in the SNAG domain of Insm1 results in impaired recruitment of Lsd1 and increased H3K4me1/2 levels at hematopoietically expressed homeobox (Hhex) loci that are bound by Insm1, thereby promoting the transcriptional activity of the δ-cell-specific gene Hhex Our study has identified a novel function of the SNAG domain of Insm1 in the regulation of pancreatic endocrine cell differentiation, particularly in the repression of β- to δ-cell transdifferentiation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-MAFB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SST antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution