MilliporeSigma
  • Diagnosing Alpha-1-Antitrypsin Deficiency Using A PCR/Luminescence-Based Technology.
Diagnosing Alpha-1-Antitrypsin Deficiency Using A PCR/Luminescence-Based Technology.
International journal of chronic obstructive pulmonary disease (2019-12-11)
Martina Veith, Andreas Klemmer, Iker Anton, Rachid El Hamss, Noelia Rapun, Sabina Janciauskiene, Viktor Kotke, Christian Herr, Robert Bals, Claus Franz Vogelmeier, Timm Greulich
ABSTRACT
Alpha-1-antitrypsin deficiency (AATD) is a rare hereditary condition resulting from the mutations in the SERPINA1 (serine protease inhibitor) gene and is characterized by low circulating levels of the alpha-1 antitrypsin (AAT) protein. The traditional algorithm for laboratory testing of AATD involves the analysis of AAT concentrations (nephelometry), phenotyping (isoelectric focusing, IEF), and genotyping (polymerase chain reaction, PCR); in selected cases, full sequencing of the SERPINA1 gene can be undertaken. New technologies arise that may make diagnosis easier and faster. We developed and evaluated a new diagnostic algorithm based on Luminex xMAP (multi-analyte profiling) technology using Progenika A1AT Genotyping Test. In an initial learning phase, 1979 samples from individuals suspected of having AATD were examined by both, a traditional and a "new" algorithm. In a second phase, 1133 samples were analyzed with the Luminex xMAP only. By introducing a Luminex xMAP based algorithm, we were able to simultaneously identify 14 mutations in SERPINA1 gene (instead of two- S and Z-by using our old algorithm). Although the quantity of IEF assays remained unchanged, the nephelometric measurements and sequencing were reduced by 79% and 63.4%, respectively. The new method is convenient, fast and user-friendly. The application of the Luminex xMAP technology can simplify and shorten the diagnostic workup of patients with suspected AATD.
Sigma-Aldrich
Neutralization Solution for Blood, sufficient for 100 reactions, Molecular Biology, sufficient for 1000 reactions