MilliporeSigma
  • Spike protein recognition of mammalian ACE2 predicts the host range and an optimized ACE2 for SARS-CoV-2 infection.
Spike protein recognition of mammalian ACE2 predicts the host range and an optimized ACE2 for SARS-CoV-2 infection.
Biochemical and biophysical research communications (2020-03-24)
Junwen Luan, Yue Lu, Xiaolu Jin, Leiliang Zhang
ABSTRACT
SARS-CoV-2 causes the recent global COVID-19 public health emergency. ACE2 is the receptor for both SARS-CoV-2 and SARS-CoV. To predict the potential host range of SARS-CoV-2, we analyzed the key residues of ACE2 for recognizing S protein. We found that most of the selected mammals including pets (dog and cat), pangolin and Circetidae mammals remained the most of key residues for association with S protein from SARS-CoV and SARS-CoV-2. The interaction interface between cat/dog/pangolin/Chinese hamster ACE2 and SARS-CoV/SARS-CoV-2 S protein was simulated through homology modeling. We identified that N82 in ACE2 showed a closer contact with SARS-CoV-2 S protein than M82 in human ACE2. Our finding will provide important insights into the host range of SARS-CoV-2 and a new strategy to design an optimized ACE2 for SARS-CoV-2 infection.
Sigma-Aldrich
SARS-COV-2-Spike-RBD epitope (370-394), ≥95% (HPLC), lyophilized powder
Sigma-Aldrich
SARS-COV-2-Spike-RBD epitope (480-499), ≥95% (HPLC), lyophilized powder
Sigma-Aldrich
Anti-SARS-COV-2-Spike-RBD region Peroxidase conjugated antibody produced in rabbit
Sigma-Aldrich
SARS-COV-2-Spike-RBD epitope (450-469), 95% (HPLC), lyophilized powder