MilliporeSigma
  • Decreased immune response in monkeys administered a human T-effector cell agonist (OX40) antibody.
Decreased immune response in monkeys administered a human T-effector cell agonist (OX40) antibody.
Toxicology and applied pharmacology (2020-10-19)
Joshua T Gamse, Wendy Freebern, Rashade Haynes Ii, Frank Simutis, Mary Pazian, James Crona, Helen G Haggerty, Michael Graziano, Roderick Todd Bunch
ABSTRACT
The OX40 receptor plays a crucial co-stimulatory role in T effector cell survival, expansion, cytokine production, and cytotoxicity to tumor cells; therefore, OX40 agonists are being evaluated as anti-cancer immunotherapies, especially in combination with checkpoint inhibitors. To support clinical development of BMS-986178 (an OX40 agonist antibody), two repeat-dose toxicity studies were conducted in cynomolgus monkeys. In the first study, BMS-986178 was administered intravenously (IV) once weekly for one month at doses from 30 to 120 mg/kg. BMS-986178 was well tolerated; surprisingly, immune function was suppressed rather than increased based on pharmacodynamic (PD) and flow cytometry readouts (e.g. T-cell dependent antibody response [TDAR]). To determine whether immune suppression was due to a bi-phasic response, a follow-up study was conducted at lower doses (1 and 10 mg/kg). Although receptor engagement was confirmed, immune function was still suppressed at both doses. In addition, treatment-emergent anti-drug antibodies (ADAs) at 1 mg/kg resulted in hypersensitivity reactions and reduced BMS-986178 exposure after repeated dosing, which precluded a full PD assessment at this dose. In conclusion, BMS-986178 was clinically well-tolerated by monkeys at weekly IV doses from 10 to 120 mg/kg (AUC[0-168] ≤ 712,000 μg●h/mL). However, despite target engagement, PD assays and other immune endpoints demonstrated immune suppression, not stimulation. Due to the inverted immune response at higher doses and the onset of ADAs, additional repeat-dose toxicity studies of BMS-986178 in monkeys (that would typically be required to support Phase 3 clinical trials and registration) would not add value for human safety assessment.
Sigma-Aldrich
Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin-Free, Sterile-Liquid, Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin Free, is a large, multi-subunit, O2-carrying, metalloprotein that is used as a carrier protein in the production of antibodies.