MilliporeSigma
  • Effects of Intensive and Semi-Intensive Production on Sheep Milk Chemical Composition, Physicochemical Characteristics, Fatty Acid Profile, and Nutritional Indices.
Effects of Intensive and Semi-Intensive Production on Sheep Milk Chemical Composition, Physicochemical Characteristics, Fatty Acid Profile, and Nutritional Indices.
Animals : an open access journal from MDPI (2021-09-29)
Eleni Kasapidou, Zoitsa Basdagianni, Vasileios Papadopoulos, Chrysoula Karaiskou, Anastasios Kesidis, Arsenios Tsiotsias
ABSTRACT
Dairy sheep farming is a vital sector of the agricultural economy in Greece. Information on the effect of the farming system on sheep milk characteristics is critical for producing dairy products with improved nutritional value according to the current guidelines for healthy food consumption. This study aimed to investigate the basic composition, physicochemical characteristics, fatty acid composition, and nutritional indices of milk produced in intensive and semi-intensive sheep farms. Bulk tank milk samples from 20 intensive and 20 semi-intensive sheep farms were examined. Sheep in the intensive farms were kept indoors and were fed with roughage, silage, and concentrates. Sheep in the semi-intensive farms were kept indoors during the night and were moved to the pastures during the day. Sheep were fed with roughage, silage, and concentrates in combination with grazing. Milk composition and somatic cell count were determined with automatic analyzers. The physicochemical characteristics were determined with standard laboratory methods. The fatty acid composition was analyzed by gas chromatography. The farming system did not affect milk chemical composition and physicochemical characteristics. However, milk fatty acid composition and nutritional value were significantly improved in milk from farms using the semi-intensive production system, and this favorable effect was attributed to the inclusion of pasture in sheep diet.
Product Description
Supelco
PUFA No.2 (Animal Source), analytical standard