MilliporeSigma
  • A gas chromatography/electron ionization-mass spectrometry-selected ion monitoring method for determining the fatty acid pattern in food after formation of fatty acid methyl esters.
A gas chromatography/electron ionization-mass spectrometry-selected ion monitoring method for determining the fatty acid pattern in food after formation of fatty acid methyl esters.
Journal of agricultural and food chemistry (2005-11-10)
Saskia Thurnhofer, Walter Vetter
ABSTRACT
A method using gas chromatography/electron ionization-mass spectrometry (GC/EI-MS) in the selected ion monitoring (SIM) mode was developed for the analysis of fatty acids as methyl esters (FAMEs) in order to determine their percentage contribution to the fatty acid profile in food. In the GC/EI-MS-SIM mode, saturated fatty acids were determined with m/z 87, monoenoic fatty acids were determined with m/z 74, and polyenoic fatty acids were determined via the sum of m/z 79 and m/z 81. The ratios of these fragment ions and the GC retention data provided additional information for tentative structural assignments. The 28 FAME standards tested provided similar results for the novel GC/EI-MS-SIM method and GC/EI-MS in the full scan mode, both of which were slightly worse than GC/flame ionization detection (FID). Analysis of sunflower oil, suet, and cod liver oil verified that both major and minor fatty acids (20-60% and down to 0.001% contribution to the fatty acid pattern) were determined with sufficient quality that justifies application of the GC/EI-MS-SIM method for the analysis of food samples. Furthermore, the method was approximately 20- or approximately 10-fold more sensitive than GC/EI-MS in the full scan mode or GC/FID, respectively. The method is suited for both quantitative purposes and fatty acid identification in samples where only low amounts of lipids are available.
Sigma-Aldrich
Methyl myristoleate, ≥98.5% (capillary GC), liquid