MilliporeSigma
  • Evidence of increased microvessel density and activation of the hypoxia pathway in tumours from the hereditary leiomyomatosis and renal cell cancer syndrome.
Evidence of increased microvessel density and activation of the hypoxia pathway in tumours from the hereditary leiomyomatosis and renal cell cancer syndrome.
The Journal of pathology (2004-12-09)
Patrick Pollard, Noel Wortham, Ella Barclay, Afrina Alam, George Elia, Sanjiv Manek, Richard Poulsom, Ian Tomlinson
ABSTRACT
The Mendelian tumour syndromes hereditary leiomyomatosis and renal cell cancer (HLRCC) and hereditary paragangliomatosis with phaeochromocytomas (HPGL) result from mutations in nuclear genes (FH and SDHB/C/D, respectively) that encode Krebs cycle enzymes. HPGL tumours are highly vascular and there is evidence that inactivation of SDH leads to activation of the hypoxia/angiogenesis pathway. In contrast, uterine leiomyomas are not generally regarded as particularly vascular lesions. In order to test the possibility that activation of the hypoxia/angiogenesis pathway contributes to tumourigenesis in HLRCC, increased vascularity and hypoxia pathway activation were searched for in HLRCC tumours. Microvessel density was markedly higher in uterine leiomyomas from HLRCC than in the surrounding myometrium; it was notable that sporadic uterine leiomyomas were actually less vascular than normal myometrium. In HLRCC tumours, there was increased expression of transcripts from the hypoxia-responsive genes vascular endothelial growth factor (VEGF) and BNIP3; sporadic uterine leiomyomas did not show these changes. All uterine leiomyomas showed decreased expression of thrombospondin 1. Although sporadic and HLRCC uterine leiomyomas appear to have identical morphology, their pathways of tumourigenesis may be fundamentally different. As is the case in HPGL, it is probable that failure of the Krebs cycle in HLRCC tumours causes inappropriate signalling that the cell is in a hypoxic state, leading to angiogenesis and perhaps directly to clonal expansion and tumour growth through some uncharacterized, cell-autonomous effect.