MilliporeSigma
Site-specific Labeling of B Cell Receptor and Soluble Immunoglobulin.
Bio-protocol (2021-03-05)
Yu Wang, Zhixun Shen, Zhengpeng Wan, Wanli Liu
ABSTRACT
B lymphocyte activation is regulated by its membrane-bound B cell receptors (BCRs) upon recognizing diverse antigens. It is hypothesized that antigen binding would trigger conformational changes within BCRs, followed by a series of downstream signaling activation. To measure the BCR conformational changes in live cells, a fluorescent site-specific labeling technique is preferred. Genetically encoded fluorescent tags visualize the location of the target proteins. However, these fluorescent proteins are large (~30 kDa) and would potentially perturb the conformation of BCRs. Here, we describe the general procedures of utilizing short tag-based site-specific labeling methodologies combining with fluorescence resonance energy transfer (FRET) assay to monitor the conformational changes within BCR extracellular domains upon antigen engagement.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder