MilliporeSigma
  • X-ray studies on crystalline complexes involving amino acids and peptides. XLII. Adipic acid complexes of L- and DL-arginine and supramolecular association in arginine-dicarboxylic acid complexes.
X-ray studies on crystalline complexes involving amino acids and peptides. XLII. Adipic acid complexes of L- and DL-arginine and supramolecular association in arginine-dicarboxylic acid complexes.
Acta crystallographica. Section B, Structural science (2005-01-22)
Siddhartha Roy, Desh Deepak Singh, M Vijayan
ABSTRACT
The adipic acid complexes of DL-arginine and L-arginine are made up of zwitterionic, singularly positively charged arginium ions and doubly negatively charged adipate ions, with a 2:1 stoichiometry. One of the two crystallographically independent arginium ions in the L-arginine complex has a conformation hitherto unobserved in crystal structures containing the amino acid. In the present study the structural data on arginine complexes of saturated dicarboxylic acids with 0-5 C atoms separating the two carboxyl functions are given. In terms of molecular aggregation, formic and acetic acid complexes behave in a similar way to those involving fairly long carboxylic acids such as adipic acid. By and large, the supramolecular assembly in complexes involving dicarboxylic acids with 3 or more C atoms separating the carboxyl groups (glutaric, adipic and pimelic acids), and those involving formic and acetic acids, have common features. The aggregation patterns in complexes involving oxalic, malonic and maleic acids do not share striking features among themselves (except for the mode of hydrogen-bonded dimerization of arginium ions) or with those involving larger dicarboxylic acids. Complexes of succinic acid, the shortest linear dicarboxylic acid, share features with those involving shorter as well as longer dicarboxylic acids. The difference in the behaviour of long and short dicarboxylic acids and the ambiguous behaviour of succinic acid can be broadly related to their lengths.
Sigma-Aldrich
DL-Arginine hydrochloride, ≥98% (TLC)