MilliporeSigma
  • Novel biodegradable copolyesters containing blocks of poly(3-hydroxyoctanoate) and poly(epsilon-caprolactone): synthesis and characterization.
Novel biodegradable copolyesters containing blocks of poly(3-hydroxyoctanoate) and poly(epsilon-caprolactone): synthesis and characterization.
Macromolecular bioscience (2004-11-13)
Laurianne Timbart, Estelle Renard, Valérie Langlois, Philippe Guerin
ABSTRACT
Novel biodegradable polyester block copolymers have been synthesized by using well-defined poly(3-hydroxyoctanoate) (PHO) oligomers having a hydroxyl end group and an ester end group with M(n) values of 800, 2,500, 5,300, 8,000, or 20,000 as an elastomeric soft segment and poly(epsilon-caprolactone) as a more crystalline segment. These PHO oligomers prepared by methanolysis were subjected to block copolymerization with epsilon-caprolactone. The chemical structure of the copolymers was confirmed by (1)H NMR and (13)C NMR spectroscopy. All the copolyesters are semi-crystalline and two T(g) were observed by differential scanning calorimetry when the molecular weight of the PHO block is about 20,000.