MilliporeSigma
  • Inhibitory effect of xylamine on the uptake of [3H]norepinephrine into primary astrocyte cultures.
Inhibitory effect of xylamine on the uptake of [3H]norepinephrine into primary astrocyte cultures.
Brain research (1992-11-27)
T K Chang, J T Cheng
ABSTRACT
Primary astrocyte cultures from neonatal rat brains show a Na+-dependent, desipramine-sensitive uptake of [3H]norepinephrine ([3H]NE). Xylamine, a nitrogen mustard, attenuated this uptake of [3H]NE into the astrocytes (IC50 = 0.8 microM). The dose-dependent reduction of [3H]NE uptake by xylamine indicated competitive kinetics. However, xylamine lost the effect if the active transport inhibitor, ouabain or iodoacetate, was also incubated during the pretreatment period of if Na+ was absent. The activity of Na(+)-K(+)-ATPase in astrocytes was not modified by xylamine at the concentrations sufficient to block the uptake of [3H]NE. These findings suggest that xylamine has the ability to compete with the transport of [3H]NE into astrocytes through an effect on the carrier instead of an action on enzymatic activity.