MilliporeSigma
  • Comparison of two alternative dominant selectable markers for wine yeast transformation.
Comparison of two alternative dominant selectable markers for wine yeast transformation.
Applied and environmental microbiology (2004-12-03)
Eduardo Cebollero, Ramon Gonzalez
ABSTRACT
Genetic improvement of industrial yeast strains is restricted by the availability of selectable transformation markers. Antibiotic resistance markers have to be avoided for public health reasons, while auxotrophy markers are generally not useful for wine yeast strain transformation because most industrial Saccharomyces cerevisiae strains are prototrophic. For this work, we performed a comparative study of the usefulness of two alternative dominant selectable markers in both episomic and centromeric plasmids. Even though the selection for sulfite resistance conferred by FZF1-4 resulted in a larger number of transformants for a laboratory strain, the p-fluoro-DL-phenylalanine resistance conferred by ARO4-OFP resulted in a more suitable selection marker for all industrial strains tested. Both episomic and centromeric constructions carrying this marker resulted in transformation frequencies close to or above 10(3) transformants per microg of DNA for the three wine yeast strains tested.
Sigma-Aldrich
p-Fluoro-DL-phenylalanine