MilliporeSigma
Preparation of Amyloid Fibril Networks.
Bio-protocol (2018-02-20)
Mirren Charnley, Jay Gilbert, Owen G Jones, Nicholas P Reynolds
ABSTRACT
Networks of amyloid nanofibrils fabricated from common globular proteins such as lysozyme and β-lactoglobulin have material properties that mimic the extracellular microenvironment of many cell types. Cells cultured on such amyloid fibril networks show improved attachment, spreading and in the case of mesenchymal stem cells improved differentiation. Here we describe a detailed protocol for fabricating amyloid fibril networks suitable for eukaryotic cell culture applications.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Pricing and availability is not currently available.
Sigma-Aldrich
Aldrich clamp holder, with zinc plated steel thumbscrews
Pricing and availability is not currently available.