MilliporeSigma
  • Overexpression of adenosine kinase in epileptic hippocampus contributes to epileptogenesis.
Overexpression of adenosine kinase in epileptic hippocampus contributes to epileptogenesis.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2004-01-23)
Nicolette Gouder, Louis Scheurer, Jean-Marc Fritschy, Detlev Boison
ABSTRACT
Endogenous adenosine in the brain is thought to prevent the development and spread of seizures via a tonic anticonvulsant effect. Brain levels of adenosine are primarily regulated by the activity of adenosine kinase. To establish a link between adenosine kinase expression and seizure activity, we analyzed the expression of adenosine kinase in the brain of control mice and in a kainic acid-induced mouse model of mesial temporal lobe epilepsy. Immunohistochemical analysis of brain sections of control mice revealed intense staining for adenosine kinase, mainly in astrocytes, which were more or less evenly distributed throughout the brain, as well as in some neurons, particularly in olfactory bulb, striatum, and brainstem. In contrast, hippocampi lesioned by a unilateral kainic acid injection displayed profound astrogliosis and therefore a significant increase in adenosine kinase immunoreactivity accompanied by a corresponding increase of enzyme activity, which paralleled chronic recurrent seizure activity in this brain region. Accordingly, seizures and interictal spikes were suppressed by the injection of a low dose of the adenosine kinase inhibitor 5-iodotubercidin. We conclude that overexpression of adenosine kinase in discrete parts of the epileptic hippocampus may contribute to the development and progression of seizure activity.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer, lyophilized powder
Sigma-Aldrich
5-Iodotubercidin, ≥85%, solid