MilliporeSigma
  • Metabolic characterization of the bovine blastocyst, inner cell mass, trophectoderm and blastocoel fluid.
Metabolic characterization of the bovine blastocyst, inner cell mass, trophectoderm and blastocoel fluid.
Reproduction (Cambridge, England) (2003-09-13)
N Gopichandran, H J Leese
ABSTRACT
The formation of a viable blastocyst is dependent upon the establishment of a correct inner cell mass (ICM):trophectoderm cell ratio but little is known about the metabolism of the two cell populations or about the composition of blastocoel fluid. In this study, the metabolism of intact bovine blastocysts, isolated ICM and trophectoderm was examined in terms of glucose and pyruvate uptake, lactate production, and amino acid consumption or production. The concentration of these nutrients in blastocoel fluid was also determined. The metabolism of glucose, pyruvate and lactate differed significantly between the isolated ICM and trophectoderm. Isolated trophectoderm had a higher pyruvate (P<0.001) and lower glucose (P<0.05) consumption, and higher lactate production (P<0.05) than did ICM. The consumption or production of amino acids by ICM and trophectoderm also differed, with the trophectoderm displaying a higher turnover (the sum of production and consumption). The ICM and trophectoderm both depleted arginine, aspartate and leucine, whereas the production of alanine was consistent. Isolated ICM depleted a further six amino acids, which appeared during trophectoderm culture; the reverse trend was observed for the remaining amino acids. The concentration of lactate in blastocoel fluid was significantly higher than in synthetic oviductal fluid supplemented with amino acids and BSA (SOFaaBSA; P<0.05). However, glucose (P<0.05) and pyruvate (P<0.001) concentrations were both lower. Aspartate, glutamate, glycine, alanine and tryptophan were present at significantly higher concentrations in blastocoel fluid than in SOFaaBSA, whereas threonine and asparagine concentrations were significantly lower. The metabolism of composite blastocysts, obtained by summing the consumption and production profiles of the ICM and trophectoderm, and taking into account their respective number of cells, was higher than that of intact blastocysts, indicating that upon isolation of the two cell populations there may be disruption to paracrine interactions or the onset of culture-induced cellular stress or both.
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)