MilliporeSigma
  • Sericin inhibits MDA‑MB‑468 cell proliferation via the PI3K/Akt pathway in triple‑negative breast cancer.
Sericin inhibits MDA‑MB‑468 cell proliferation via the PI3K/Akt pathway in triple‑negative breast cancer.
Molecular medicine reports (2020-12-15)
Lin Niu, Songhe Yang, Xueying Zhao, Xiaochao Liu, Lina Si, Meng Wei, Lei Liu, Luyang Cheng, Yuebing Qiao, Zhihong Chen
ABSTRACT
Triple negative breast cancer (TNBC) is a subtype of breast cancer characterized by an aggressive histology and poor prognosis, with limited treatment options in the clinic. In the present study, the effect of sericin, as an anti‑cancer drug, on TNBC cell proliferation was investigated using a MTT assay, a colony formation assay and immunocytochemistry staining of Ki67. Results from the flow cytometry demonstrated that sericin induced G0/G1 cell cycle arrest and promoted cellular apoptosis. Cell cycle and apoptosis‑related proteins were detected via western blot analysis. Immunocytochemistry staining identified that P21 was translocated into the nucleus. Additionally, several pathways were significantly enriched in TNBC based on the Gene Expression Omnibus database, with the most prominent pathway being the PI3K/Akt signaling pathway. In TNBC MDA‑MB‑468 cells, sericin suppressed the PI3K/Akt pathway. All these findings suggested that sericin served a critical role in suppressing TNBC cell proliferation, inducing cell cycle arrest and promoting cellular apoptosis. The results indicated that the underlying molecular mechanism was, at least partially, via the downregulation of the PI3K/Akt signaling pathway.
Sigma-Aldrich
Sericin Bombyx mori (silkworm)