MilliporeSigma
  • Evaluation of Small Molecule Combinations against Respiratory Syncytial Virus In Vitro.
Evaluation of Small Molecule Combinations against Respiratory Syncytial Virus In Vitro.
Molecules (Basel, Switzerland) (2021-05-06)
Yuzhen Gao, Jingjing Cao, Pan Xing, Ralf Altmeyer, Youming Zhang
ABSTRACT
Respiratory syncytial virus (RSV) is a major pathogen that causes severe lower respiratory tract infection in infants, the elderly and the immunocompromised worldwide. At present no approved specific drugs or vaccines are available to treat this pathogen. Recently, several promising candidates targeting RSV entry and multiplication steps are under investigation. However, it is possible to lead to drug resistance under the long-term treatment. Therapeutic combinations constitute an alternative to prevent resistance and reduce antiviral doses. Therefore, we tested in vitro two-drug combinations of fusion inhibitors (GS5806, Ziresovir and BMS433771) and RNA-dependent RNA polymerase complex (RdRp) inhibitors (ALS8176, RSV604, and Cyclopamine). The statistical program MacSynergy II was employed to determine synergism, additivity or antagonism between drugs. From the result, we found that combinations of ALS8176 and Ziresovir or GS5806 exhibit additive effects against RSV in vitro, with interaction volume of 50 µM2% and 31 µM2% at 95% confidence interval, respectively. On the other hand, all combinations between fusion inhibitors showed antagonistic effects against RSV in vitro, with volume of antagonism ranging from -50 µM2 % to -176 µM2 % at 95% confidence interval. Over all, our results suggest the potentially therapeutic combinations in combating RSV in vitro could be considered for further animal and clinical evaluations.
Sigma-Aldrich
RSV604, ≥98% (HPLC)