MilliporeSigma
  • Extracellular Concentration of L-Cystine Determines the Sensitivity to System xc - Inhibitors.
Extracellular Concentration of L-Cystine Determines the Sensitivity to System xc - Inhibitors.
Biomolecules & therapeutics (2021-10-21)
Md Abdullah, Seung Jin Lee
ABSTRACT
Targeting the cystine/glutamate exchange transporter, system xc -, is a promising anticancer strategy that induces ferroptosis, which is a distinct form of cell death mediated by iron-dependent lipid peroxidation. The concentration of L-cystine in culture medium is higher than the physiological level. This study was aimed to evaluate the effects of L-cystine concentration on the efficacy of ferroptosis inducers in hepatocellular carcinoma cells. This study showed that treatment with sulfasalazine or erastin, a system xc - inhibitor, decreased the viability of Huh6 and Huh7 cells in a dose-dependent manner, and the degree of growth inhibition was greater in medium containing a physiological L-cystine concentration of 83 μM than in commercial medium with a concentration of 200 μM L-cystine. However, RSL3, a glutathione peroxidase 4 inhibitor, decreased cell viability to a similar extent in media containing both L-cystine concentrations. Sulfasalazine and erastin significantly increased the percentages of propidium iodide-positive cells in media with 83 μM L-cystine, but not in media with 200 μM L-cystine. Sulfasalazine- or erastin-induced accumulation of lipid peroxidation as monitored by C11-BODIPY probe was higher in media with 83 μM L-cystine than in media with 200 μM L-cystine. In contrast, the changes in the percentages of propidium iodide-positive cells and lipid peroxidation by RSL3 were similar in both media. These results showed that sulfasalazine and erastin, but not RSL3, were efficacious under conditions of physiological Lcystine concentration, suggesting that medium conditions would be crucial for the design of a bioassay for system xc - inhibitors.
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.