MilliporeSigma
  • Measurement in saliva from neurotypical adults of biomarkers pertinent to autism spectrum disorders.
Measurement in saliva from neurotypical adults of biomarkers pertinent to autism spectrum disorders.
Future science OA (2016-12-30)
Helen V Ratajczak, Robert B Sothern
ABSTRACT
Measure biomarkers pertinent to autism in saliva from humans. At 7:30 PM (reading instructions) and 8:30 PM (hearing instructions), neurotypical adults (6 M, 6 F) each spat into tubes containing protease inhibitors. Cells were counted, samples aliquoted, frozen and thawed. Rationale was given for choice of biomarkers. ELISA: CD26, IL-12, carnitine, C4B, GSH, GSSG, MT-2, testosterone, IFN-γ. Mass spectrometry: cystine, glutamine, glutamic acid, GABA, serotonin. Electrochemiluminescentimmunoassay: cortisol. Radioimmunoassay: melatonin. Cells averaged 2.16 × 106/ml. M > F: CD-26, C4B, MT-2. Testosterone, cortisol. Glutamine, glutamic acid, IFN-γ, melatonin and GSSG were measurable. Remaining biomarkers were measured in <50% of samples. Concentrations were equal at both times. Saliva can be collected by literate individuals without added instruction. Ten biomarkers were measurable.
Product Description
Supelco
Cortisol-D4 (9,11,12, 12-D4) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Serotonin hydrochloride, powder