MilliporeSigma
  • Valorization of lignin components into gallate by integrated biological hydroxylation, O-demethylation, and aryl side-chain oxidation.
Valorization of lignin components into gallate by integrated biological hydroxylation, O-demethylation, and aryl side-chain oxidation.
Science advances (2021-09-14)
Chenggu Cai, Zhaoxian Xu, Huarong Zhou, Sitong Chen, Mingjie Jin
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Hoagland′s No. 2 Basal Salt Mixture, powder, suitable for plant cell culture