MilliporeSigma
  • Fabrication of Photothermo-Responsive Drug-Loaded Nanogel for Synergetic Cancer Therapy.
Fabrication of Photothermo-Responsive Drug-Loaded Nanogel for Synergetic Cancer Therapy.
Polymers (2019-04-10)
Ray Chang, Wei-Bor Tsai
ABSTRACT
Temperature stimulus, easy modulation in comparison to other environmental stimuli, makes thermo-responsive nanocarriers popular in the applications of controlled drug release for cancer therapy. In this study, photosensitive sodium copper chlorophyllin (SCC) was incorporated into thermo-responsive polymeric nanogels consisted of N-isopropylacrylamide and N-(hydroxymethyl)acrylamide. Significant heat was generated from the SCC-containing nanogels under the exposure to 532-nm green laser, and resulted in cell mortality. The thermo-responsive nanogel loaded with 5-FU, an anti-cancer drug, released the drug explosively when exposed to green laser. The combination of hyperthermia and temperature-induced drug release via green laser irradiation greatly enhanced cell mortality to a maximal extent. Such photothermo-responsive nanogel possesses a great potential in anti-cancer treatment.
Sigma-Aldrich
4,4′-Azobis(4-cyanovaleric acid), ≥98.0% (T)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%