MilliporeSigma
  • Characterization and comparison of the digestive physiology of two scombrids, Katsuwonus pelamis and Sarda sarda, in the Gulf of Cádiz.
Characterization and comparison of the digestive physiology of two scombrids, Katsuwonus pelamis and Sarda sarda, in the Gulf of Cádiz.
PloS one (2021-04-15)
Diogo Dias, Gian Marco Dardengo, Sofia Engrola, Carmen Navarro-Guillén
ABSTRACT
Fish and other marine animals have a unique and intimate interaction with their surrounding environment. Diet type or trophic level play significant roles in modulating species digestive physiology. However, little is known about how the trophic niche influences digestive activity and gut microbiota in scombrids species. The aim of the present study was to analyse and describe the digestive physiology of the skipjack tuna (Katsuwonus pelamis) and the Atlantic bonito (Sarda sarda) as bioindicator of the feeding ecology and trophic niche for both species in the Gulf of Cádiz (Spain). For that, fish proximate composition, pH of digestive organs and digestive enzymes activity levels were analysed in 10 individuals of each species to gain insight into the digestive physiological adaptations of the two species of scombrids. In addition, intestinal microbiota composition was determined for the skipjack tuna. The integration of the results suggested a clear trophic niche segregation between both species. Stomach pH was associated in both species with infrequent feeding events. Body proximate composition and digestive lipases activity patterns pointed to a high predominance of lipids in the Atlantic bonito diet, suggesting oily fish as main prey. On the other hand, results supported the skipjack tuna as a highly opportunistic feeder with a more varied diet, including fish but also crustaceans as preys. The gut microbial community in the latter species is dominated by Firmicutes and Tenericutes at phylum level, and by Mycoplasma, Blautia and Dorea at genus level. The present study contributes to advance the knowledge on the feeding behaviour and physiology diversity in fish species as a result of adaptation to a particular habitat.
Sigma-Aldrich
4-Methylumbelliferyl N-acetyl-β-D-glucosaminide, suitable for fluorescence, ≥99.0% (TLC)